Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppp2r5ea | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194415.2 | NP_919396.1 | 13 - 14 | 1760 | 65 | |
GenBank mRNA | AY150214.1 | - | 12 - 13 | 1307 | 65 | |
BC044398.1 | - | 13 - 14 | 1576 | 65 | ||
BC078645.1 | - | 13 - 14 | 1760 | 65 | ||
BC165347.1 | - | 12 - 13 | 1341 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |