Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: setd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199600.1 | NP_955894.1 | 3 - 4 | 422 | 67 | |
GenBank mRNA | BC044440.1 | - | 3 - 4 | 438 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |