Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: setd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199600.1 | NP_955894.1 | 6 - 7 | 889 | 92 | |
GenBank mRNA | BC044440.1 | - | 6 - 7 | 905 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |