Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199691.2 | NP_955985.2 | 6 - 7 | 1038 | 82 | |
GenBank mRNA | AY423034.1 | - | 6 - 7 | 1031 | 82 | |
BC060901.1 | - | 6 - 7 | 980 | 82 | ||
BC065357.1 | - | 6 - 7 | 1000 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |