Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199691.2 | NP_955985.2 | 7 - 8 | 1179 | 86 | |
GenBank mRNA | AY423034.1 | - | 7 - 8 | 1172 | 86 | |
BC060901.1 | - | 7 - 8 | 1121 | 86 | ||
BC065357.1 | - | 7 - 8 | 1141 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |