Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200238.1 | NP_956532.1 | 12 - 13 | 1428 | 126 | |
XM_005167920.4 | XP_005167977.1 | 12 - 13 | 1506 | 126 | ||
GenBank mRNA | BC047186.1 | - | 12 - 13 | 1428 | 126 | |
BC165692.1 | - | 11 - 12 | 1381 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |