Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200238.1 | NP_956532.1 | 8 - 9 | 934 | 105 | |
XM_005167920.4 | XP_005167977.1 | 8 - 9 | 1012 | 105 | ||
GenBank mRNA | BC047186.1 | - | 8 - 9 | 934 | 105 | |
BC165692.1 | - | 7 - 8 | 887 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |