Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcolce2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200460.1 | NP_956754.1 | 6 - 7 | 926 | 57 | |
GenBank mRNA | BC055153.1 | - | 6 - 7 | 926 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |