Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mboat7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200537.1 | NP_956831.1 | 3 - 4 | 277 | 89 | |
XM_021466647.1 | XP_021322322.1 | 3 - 4 | 372 | 89 | ||
GenBank mRNA | BC056305.1 | - | 3 - 4 | 277 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |