Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tsc22d3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200569.2 | NP_956863.1 | 1 - 2 | 296 | 113 | |
GenBank mRNA | BC056586.1 | - | 1 - 2 | 284 | 113 | |
BC068985.1 | - | 1 - 2 | 280 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |