Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trip13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200582.1 | NP_956876.1 | 10 - 11 | 891 | 63 | |
XM_005170125.4 | XP_005170182.1 | 10 - 11 | 63 | |||
XM_017351326.2 | XP_017206815.1 | 10 - 11 | 63 | |||
GenBank mRNA | BC056713.1 | - | 10 - 11 | 891 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |