Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cacng2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200641.1 | NP_956935.1 | 3 - 4 | 925 | 97 | |
GenBank mRNA | BC057439.1 | - | 3 - 4 | 925 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |