Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ssu72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200728.1 | NP_957022.1 | 2 - 3 | 287 | 66 | |
GenBank mRNA | BC059504.1 | - | 2 - 3 | 287 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |