Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rabgef1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200941.1 | NP_957235.1 | 4 - 5 | 611 | 65 | |
GenBank mRNA | BC045469.1 | - | 4 - 5 | 611 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |