Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sh2d3cb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201076.1 | NP_957370.1 | 10 - 11 | 1962 | 66 | |
XM_005165604.4 | XP_005165661.1 | 10 - 11 | 1899 | 66 | ||
XM_021476041.1 | XP_021331716.1 | 10 - 11 | 1884 | 66 | ||
GenBank mRNA | BC049321.1 | - | 10 - 11 | 1962 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |