Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201153.1 | NP_957447.1 | 2 - 3 | 190 | 75 | |
GenBank mRNA | AY394937.1 | - | 2 - 3 | 164 | 75 | |
BC045902.1 | - | 2 - 3 | 190 | 75 | ||
BC165099.1 | - | 2 - 3 | 197 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |