Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201197.1 | NP_957491.1 | 3 - 4 | 468 | 60 | |
XM_005167473.2 | XP_005167530.1 | 3 - 4 | 1100 | 60 | ||
XM_005167474.4 | XP_005167531.1 | 4 - 5 | 744 | 60 | ||
GenBank mRNA | BC049051.1 | - | 3 - 4 | 468 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |