Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tra2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201197.1 | NP_957491.1 | 7 - 8 | 948 | 120 | |
XM_005167473.2 | XP_005167530.1 | 7 - 8 | 1580 | 120 | ||
XM_005167474.4 | XP_005167531.1 | 8 - 9 | 1224 | 120 | ||
GenBank mRNA | BC049051.1 | - | 7 - 8 | 948 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |