Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prpf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201467.1 | NP_958875.1 | 7 - 8 | 688 | 64 | |
GenBank mRNA | BC045954.1 | - | 7 - 8 | 688 | 64 | |
BC165225.1 | - | 7 - 8 | 604 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |