Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: setb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201468.1 | NP_958876.1 | 3 - 4 | 485 | 64 | |
XM_005161091.4 | XP_005161148.1 | 3 - 4 | 503 | 64 | ||
GenBank mRNA | AY398416.1 | - | 3 - 4 | 485 | 64 | |
BC066410.1 | - | 3 - 4 | 439 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |