Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dcun1d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205538.1 | NP_991101.1 | 3 - 4 | 411 | 87 | |
XM_005167810.4 | XP_005167867.1 | 3 - 4 | 516 | 87 | ||
XM_005167811.4 | XP_005167868.1 | 3 - 4 | 538 | 87 | ||
XM_021478598.1 | XP_021334273.1 | 3 - 4 | 483 | 87 | ||
GenBank mRNA | BC065884.1 | - | 3 - 4 | 411 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |