Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnph1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212589.2 | NP_997754.2 | 6 - 7 | 811 | 130 | |
XM_021468753.1 | XP_021324428.1 | 5 - 6 | 707 | 130 | ||
GenBank mRNA | BC044161.1 | - | 6 - 7 | 804 | 130 | |
BC067720.1 | - | 4 - 5 | 532 | 130 | ||
BC164507.1 | - | 5 - 6 | 736 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |