Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnph1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212589.2 | NP_997754.2 | 9 - 10 | 1144 | 61 | |
XM_021468753.1 | XP_021324428.1 | 8 - 9 | 1040 | 61 | ||
GenBank mRNA | BC044161.1 | - | 9 - 10 | 1137 | 61 | |
BC067720.1 | - | 7 - 8 | 865 | 61 | ||
BC164507.1 | - | 8 - 9 | 1069 | 61 | ||
CT671502.2 | - | 4 - 5 | 418 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |