Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptp4a3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213181.1 | NP_998346.1 | 4 - 5 | 887 | 97 | |
XM_005170023.1 | XP_005170080.1 | 5 - 6 | 97 | |||
XM_005170024.4 | XP_005170081.1 | 5 - 6 | 97 | |||
GenBank mRNA | BC067186.1 | - | 4 - 5 | 887 | 97 | |
BC152190.1 | - | 4 - 5 | 521 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |