Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnpdl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_137312.1 | 7 - 8 | 1051 | 62 | ||
XM_021479149.1 | XP_021334824.1 | 7 - 8 | 1056 | 62 | ||
GenBank mRNA | BC055525.1 | - | 7 - 8 | 1051 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |