Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213428.2 | NP_998593.1 | 5 - 6 | 556 | 92 | |
XM_005164484.2 | XP_005164541.1 | 6 - 7 | 752 | 92 | ||
GenBank mRNA | BC049036.1 | - | 5 - 6 | 557 | 92 | |
BC071429.1 | - | 5 - 6 | 534 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |