Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: setd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002456.1 | NP_001002456.1 | 2 - 3 | 257 | 78 | |
GenBank mRNA | BC076014.1 | - | 2 - 3 | 257 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |