Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcl9l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005575.1 | NP_001005575.1 | 7 - 8 | 3520 | 99 | |
XM_005159238.4 | XP_005159295.1 | 7 - 8 | 3627 | 99 | ||
XM_005159239.4 | XP_005159296.1 | 7 - 8 | 3625 | 99 | ||
GenBank mRNA | AY296057.1 | - | 7 - 8 | 3520 | 99 | |
BC163544.1 | - | 7 - 8 | 3503 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |