Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpa1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007066.2 | NP_001007067.1 | 14 - 15 | 1756 | 121 | |
XM_009297331.3 | XP_009295606.1 | 14 - 15 | 2082 | 121 | ||
XM_021470156.1 | XP_021325831.1 | 14 - 15 | 2099 | 121 | ||
GenBank mRNA | AY677197.1 | - | 14 - 15 | 1761 | 121 | |
EU826642.1 | - | 13 - 14 | 1634 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |