Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc35c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008590.1 | NP_001008590.1 | 2 - 3 | 633 | 56 | |
XM_005169053.4 | XP_005169110.1 | 1 - 2 | 763 | 56 | ||
GenBank mRNA | BC086750.1 | - | 2 - 3 | 633 | 56 | |
BC154758.1 | - | 2 - 3 | 560 | 56 | ||
BC164162.1 | - | 1 - 2 | 509 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |