Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ogna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013570.2 | NP_001013588.1 | 1 - 2 | 247 | 102 | |
GenBank mRNA | BC090507.1 | - | 1 - 2 | 251 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |