Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017711.2 | NP_001017711.2 | 5 - 6 | 1292 | 110 | |
GenBank mRNA | AB212994.1 | - | 5 - 6 | 1292 | 110 | |
AY900003.1 | - | 4 - 5 | 1116 | 110 | ||
BC093257.1 | - | 5 - 6 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |