Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syt5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020546.2 | NP_001018382.2 | 8 - 9 | 1146 | 79 | |
GenBank mRNA | BC095109.1 | - | 8 - 9 | 1146 | 79 | |
BC165563.1 | - | 7 - 8 | 1063 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |