Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acbd5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020647.1 | NP_001018483.1 | 8 - 9 | 972 | 62 | |
XM_005162509.4 | XP_005162566.1 | 8 - 9 | 994 | 62 | ||
XM_009297238.3 | XP_009295513.1 | 9 - 10 | 1027 | 62 | ||
XM_009297239.3 | XP_009295514.1 | 9 - 10 | 1012 | 62 | ||
XM_009297241.3 | XP_009295516.1 | 9 - 10 | 1069 | 62 | ||
XM_021470173.1 | XP_021325848.1 | 9 - 10 | 1169 | 62 | ||
XM_021470174.1 | XP_021325849.1 | 9 - 10 | 1024 | 62 | ||
GenBank mRNA | BC095655.1 | - | 8 - 9 | 972 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |