Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lpgat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029958.1 | NP_001025129.1 | 2 - 3 | 295 | 71 | |
XM_005160439.4 | XP_005160496.1 | 2 - 3 | 707 | 71 | ||
GenBank mRNA | BC098616.1 | - | 2 - 3 | 295 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |