Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fstl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030111.2 | NP_001025282.2 | 4 - 5 | 805 | 120 | |
GenBank mRNA | BC097220.1 | - | 4 - 5 | 780 | 120 | |
BC134879.1 | - | 4 - 5 | 805 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |