Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab27a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030257.1 | NP_001025428.1 | 3 - 4 | 441 | 90 | |
GenBank mRNA | BC097191.1 | - | 3 - 4 | 441 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |