Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031674.2 | NP_001026844.2 | 2 - 3 | 806 | 79 | |
GenBank mRNA | BC114224.1 | - | 2 - 3 | 806 | 79 | |
DQ122929.1 | - | 2 - 3 | 810 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |