Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rtn4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040335.1 | NP_001035425.1 | 2 - 3 | 1292 | 92 | |
GenBank mRNA | AY555051.1 | - | 2 - 3 | 1292 | 92 | |
AY555052.1 | - | 3 - 4 | 789 | 92 | ||
AY555053.1 | - | 2 - 3 | 1008 | 92 | ||
AY555054.1 | - | 2 - 3 | 1008 | 92 | ||
AY555055.1 | - | 2 - 3 | 1008 | 92 | ||
AY555056.1 | - | 2 - 3 | 1008 | 92 | ||
AY555057.1 | - | 2 - 3 | 1008 | 92 | ||
AY555058.1 | - | 2 - 3 | 1008 | 92 | ||
AY555059.1 | - | 2 - 3 | 1008 | 92 | ||
AY555060.1 | - | 2 - 3 | 1008 | 92 | ||
AY555061.1 | - | 2 - 3 | 1008 | 92 | ||
BC068434.1 | - | 2 - 3 | 1154 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |