Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hrh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042731.1 | NP_001036196.1 | 1 | 1551 | 67 | |
XM_009305727.3 | XP_009304002.1 | 2 | 1764 | 67 | ||
GenBank mRNA | BC162414.1 | - | 1 | 1635 | 67 | |
DQ647806.1 | - | 1 | 1551 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |