Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prtga | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045030.1 | 410 | 84 | |||
GenBank mRNA | BC162624.1 | - | 380 | 84 | ||
BC162639.1 | - | 380 | 84 | |||
DQ360115.1 | - | 410 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio |