Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crb2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045162.1 | NP_001038627.1 | 8 - 9 | 2533 | 70 | |
XM_017357360.2 | XP_017212849.1 | 8 - 9 | 2575 | 70 | ||
GenBank mRNA | DQ314737.1 | - | 8 - 9 | 2533 | 70 | |
JQ740885.1 | - | 2 - 3 | 1165 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |