Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: blvra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076601.1 | NP_001070069.1 | 4 - 5 | 452 | 62 | |
GenBank mRNA | BC124225.1 | - | 4 - 5 | 452 | 62 | |
BC163949.1 | - | 3 - 4 | 370 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |