Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: me1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC124633.1 | - | 9 - 10 | 1067 | 77 |
Gene Symbol: LOC101882012 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005174538.3 | XP_005174595.1 | 4 - 5 | 408 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |