Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079980.1 | NP_001073449.1 | 4 - 5 | 203 | 153 | |
XM_005159923.4 | XP_005159980.1 | 4 - 5 | 287 | 153 | ||
XM_005159924.4 | XP_005159981.1 | 4 - 5 | 287 | 111 | ||
XM_005159925.4 | XP_005159982.1 | 4 - 5 | 261 | 111 | ||
GenBank mRNA | BC125914.1 | - | 4 - 5 | 203 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |