Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptprfb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083576.1 | NP_001077045.1 | 5 - 6 | 945 | 66 | |
XM_009298461.3 | XP_009296736.1 | 5 - 6 | 993 | 66 | ||
XM_009298464.3 | XP_009296739.1 | 5 - 6 | 993 | 66 | ||
XM_009298466.3 | XP_009296741.1 | 5 - 6 | 992 | 66 | ||
XM_009298467.3 | XP_009296742.1 | 5 - 6 | 992 | 66 | ||
XM_009298469.3 | XP_009296744.1 | 5 - 6 | 992 | 66 | ||
XM_021481419.1 | XP_021337094.1 | 5 - 6 | 993 | 66 | ||
XM_021481438.1 | XP_021337113.1 | 5 - 6 | 992 | 66 | ||
XM_021481440.1 | XP_021337115.1 | 5 - 6 | 992 | 66 | ||
XM_021481443.1 | XP_021337118.1 | 5 - 6 | 992 | 66 | ||
GenBank mRNA | BC134804.1 | - | 5 - 6 | 945 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |