Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dph2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089397.1 | NP_001082866.1 | 6 - 7 | 1395 | 122 | |
GenBank mRNA | BC134939.1 | - | 6 - 7 | 1395 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |