Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rock2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001093747.2 | NP_001087216.1 | 20 - 21 | 2580 | 106 | |
XM_009294695.3 | XP_009292970.1 | 20 - 21 | 3052 | 106 | ||
XM_009294697.3 | XP_009292972.1 | 8 - 9 | 1118 | 106 | ||
XM_017352433.2 | XP_017207922.1 | 20 - 21 | 3053 | 106 | ||
XM_017352434.2 | XP_017207923.1 | 20 - 21 | 3053 | 106 | ||
XM_017352437.2 | XP_017207926.1 | 20 - 21 | 3056 | 106 | ||
XM_021468356.1 | XP_021324031.1 | 20 - 21 | 3055 | 106 | ||
XM_021468357.1 | XP_021324032.1 | 20 - 21 | 3054 | 106 | ||
XM_021468358.1 | XP_021324033.1 | 20 - 21 | 3054 | 106 | ||
XM_021468359.1 | XP_021324034.1 | 20 - 21 | 3054 | 106 | ||
XM_021468360.1 | XP_021324035.1 | 20 - 21 | 3057 | 106 | ||
XM_021468361.1 | XP_021324036.1 | 20 - 21 | 3055 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |