Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rims3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100009.2 | NP_001093479.1 | 3 - 4 | 467 | 100 | |
XM_005170052.4 | XP_005170109.1 | 4 - 5 | 745 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |