Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130914.2 | NP_570989.2 | 4 - 5 | 642 | 66 | |
GenBank mRNA | AF179406.1 | - | 4 - 5 | 642 | 66 | |
BC162554.1 | - | 4 - 5 | 592 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |